Avantor® Hichrom, PEEK Tubing
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
PEEK tubing with precision bore for LC, LCMS/automation usage.
- Chemical resistant/inert to solvents
Ensures reproducibility in chromatographic flow rates through smooth internal surface with tight toleranced.
Customers Who Bought This Also Bought