Avantor® Hichrom, PEEK Frit
# de catalogue 76399-142
Fournisseur: HICHROM LIMITED
Spécifications
- Porosity:2.0 µm
- Description:Frit
- Frit Material:PEEK
Spécifications
A propos de cet article
Customers Who Bought This Also Bought
VWR® Snap Caps for 11 mm Snap Ring Vials, Wide Opening
Fournisseur: VWR International
81,56 $ to 628,06 $
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Fournisseur: VWR International
881,80 $ to 2 807,56 $
VWR® 11 mm Crimp Top Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
45,90 $ to 336,74 $
Screw-Thread Caps, 9 mm, MicroLiter, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
88,41 $
WHEATON® MicroLiter Crimp-Top µLVials™, Component Kits, 11 mm, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
253,80 $
SureSTART™ Caps for 2 ml Glass Snap Top Vials, Level 2 High-Throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
461,12 $ to 461,12 $
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
149,40 $
WHEATON® E-Z Vial® Large-Opening Crimp-Top Vials, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
70,19 $
WHEATON® ABC Vial™ Large-Opening Screw-Thread Vials, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
99,27 $ to 725,06 $
WHEATON® MicroLiter 20 mm Crimp Seals with Septa, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
160,41 $