Avantor® Hichrom, Connectors, Tees and Crosses, PEEK
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Use with ¹/₁₆" outer Ø PEEK™ or stainless steel tubing.
- Maximum operating pressure 241 bar (3500 psi)
Fits tubing 0.5×1.58 mm (I.D.×O.D.)
Delivery: Includes 3 or 4 (76399-380) fingertight fittings.
Customers Who Bought This Also Bought