Avantor® Hichrom, Tubing Cutter for Polymer Tubing
# de catalogue 76399-188
Fournisseur: HICHROM LIMITED
Spécifications
- Description:Standard polymer tubing cutter
- Cat. no.:76399-188
Spécifications
A propos de cet article
Standard cutter for 1/16" and 1/8" O.D. tubing.
- Compatible with PEEK and PTFE tubing
- Flat burr-free cuts
- Guide holes to ensure precise cutting
Delivery: Includes five replacement blades.
Customers Who Bought This Also Bought