Avantor® Hichrom, End Fittings
Fournisseur: HICHROM LIMITED
Certificats
Customers Who Bought This Also Bought
VWR® Snap Caps for 11 mm Snap Ring Vials, Wide Opening
Fournisseur: VWR International
$73.75 to $568.62
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$118.61 to $1,128.11
WHEATON® MicroLiter 11 mm Crimp-Top Seals, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$79.24
WHEATON® MicroLiter 9 mm Screw-Thread Vials, Component Kits, Unassembled, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$133.66