Avantor® Hichrom, PEEK Tubing
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Precision bore PEEK tubing is designed specifically for modern LC, LCMS and automation usage.
- Chemical resistant and inert to most commonly used solvents
PEEK tubing is a flexible alternative to stainless steel tubing in high pressure applications. Also ensures reproducibility in chromatographic flow rates through smooth internal surface along with consistent and tight toleranced IDs.
Customers Who Bought This Also Bought