Avantor® Hichrom, Flangeless Ferrules
Fournisseur: HICHROM LIMITED
Certificats
Certificats
A propos de cet article
Customers Who Bought This Also Bought
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$138.51
RSA™ and RSA-Pro™ Autosampler Vials, Caps and Convenient Kits
Fournisseur: MicroSolv Technology Corp
$433.35
SureSTART™ Screw Caps, 9 mm, Level 2 High-throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$2,443.19 to $2,443.19
SureSTART™ Polypropylene Screw Top 2 ml Microvials for <2 ml Samples, Level 1 Everyday Analysis, Thermo Scientific
Fournisseur: Thermo Scientific
$390.18 to $390.18
SureSTART™ Crimp Caps, 20 mm, Level 2 High-throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$180.01 to $182.41