Polygosil 100-10, HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
Certificats
A propos de cet article
Irregular silica phases for analytical applications.
- pH stability: 2 to 8
Column hardware: Stainless steel.
Spécifications
- Catalog No:
- 76402-308
- Surface area:
- 280 m²/g
Spécifications
Customers Who Bought This Also Bought
AQ™ Advanced Quality Borosilicate Glass and Plastic Inserts
Fournisseur: MicroSolv Technology Corp
$510.94
SureSTART™ Screw Caps, 9 mm, Level 3 High Performance Applications
Fournisseur: Thermo Scientific
$79.21 to $82.14
AQR™ Caps for RSA™ Autosampler Vials, MicroSolv Technology Corporation
Fournisseur: MicroSolv Technology Corp
$54.30
WHEATON® MicroLiter Screw-Thread Vials, Component Kits, Assembled, 9 mm, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$127.72
RSA™ and RSA-Pro™ Autosampler Vials, Caps and Convenient Kits
Fournisseur: MicroSolv Technology Corp
$438.81
SureSTART™ Glass Crimp Top 0.3 ml Microvials for <2 ml Samples, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$9,267.32 to $9,267.32
VWR® 11 mm Crimp Top Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$279.61
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$149.40
WHEATON® MicroLiter Crimp-Top Vials, 11 mm,Component Kits, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$142.31