Avantor® Hichrom, Cutters, Polymeric Tubing
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
The Clean-cut™ cutter is designed specifically for cutting polymeric HPLC tubing from ¹/₁₆ to ¹/₈" with no distortion of cross-section and without closing the bore.
- Cuts PEEK and PTFE tubing
- Square, burr-free cuts
- Convenient, replaceable blade
The 'Guillotine' cutter is an alternative design for cutting polymeric tubing of ¹/₁₆ to ¹/₂". A razor-sharp blade cuts while a V-shaped grove on the handle holds the tubing at the right angle to produce a square cut.
Customers Who Bought This Also Bought