Avantor® Hichrom, In-Line Filter
# de catalogue 76399-690
Fournisseur: HICHROM LIMITED
Spécifications
- Description:Fingertight Ti PEEK encased inline filter
- Cat. no.:76399-690
Spécifications
A propos de cet article
This is a fingertight bioanalytical inline filter, comprising a PEEK holder and a 5 μm titanium PEEK-encased frit.
- Protects the injector from particulates
- Compatible with all HPLC systems
- Replacement 5 μm and 2 μm titanium PEEK-encased frits are available
Placed between the pump and sample injection valve, the in-line filter traps particles released through normal piston seal wear within the pump. It is recommended that the frits are checked frequently and replaced as soon as they contribute to a system back pressure.
Customers Who Bought This Also Bought