Avantor® Hichrom PEEKsil™, Tubing
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
PEEKsil™ tubing is mechanically strong and has ideal characteristics for sealing with polymer fittings. It is comprised of high quality fused silica sheathed by PEEK tubing giving it excellent chemical compatibility.
Customers Who Bought This Also Bought