Avantor® Hichrom Anion/S, Anion Exchange HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Avantor® Hichrom Anion/S columns are based on 10 µm silica with quaternary ammonium ion-exchange groups.
- Silica-based for symmetrical peak shapes
- Separates inorganic and organic anions
These columns are optimized for non-suppressed conductivity detection and are best suited for routine separations of chloride, bromide, nitrate and sulfate.
BodyTextAccessories: Use guard cartridges with holder (75830-798) and coupler (75830-622).
Customers Who Bought This Also Bought