Avantor® Hichrom Bottom-of-the-Bottle™, Inlet Solvent Filters
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
These filters are made from 100% PEEK.
- Biocompatible filters
- Two built-in PEEK frits
The bottom frit (2 or 10 µm) will draw solvents from within 2 mm of the bottom of the solvent bottle. The 2 μm frit on the side may be used for a 203 mm Ext.Ø helium sparging line.
Customers Who Bought This Also Bought