Avantor® Hichrom, ValvTool
# de catalogue 76399-202
Fournisseur: HICHROM LIMITED
Spécifications
- Description:ValvTool
- Cat. no.:76399-202
Spécifications
A propos de cet article
This Valvtool is a uniquely designed slotted wrench which provides easy access to many hard-to-reach areas.
It enables fittings, sample loops and most other ¹/₄" stainless steel and PEEK HPLC fittings to be more easily tightened or loosened.
Customers Who Bought This Also Bought