Exsil C1, HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Exsil HPLC columns are a useful alternative to the classical spherical HPLC silicas.
- Spherical porous silica
- Competitively priced
- High efficiency
Exsil 100, the standard material, is supplied in a range of particle sizes and bonded phases to cover a wide range of applications. Exsil 80 is a higher surface area, complementary range of materials.
Spécifications
- Catalog No:
- 76398-658
- Carbon load:
- 3%
- Endcapped:
- No
- Surface area:
- 200 m²/g