Avantor® Hichrom, Tubing Cutters, Stainless Steel
Fournisseur: HICHROM LIMITED
Certificats
Certificats
A propos de cet article
Stainless steel cutters are specifically designed for ¹/₁₆" stainless steel tubing.
- Smooth and uniform cuts
Customers Who Bought This Also Bought
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$63.72 to $462.44
WHEATON® MicroLiter Septa for 20 mm Crimp Seals, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$343.64
SureSTART™ Glass Inserts for 2 ml Vials, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$120.60 to $125.07
WHEATON® ABC Vial™ Large-Opening Screw-Thread Vials, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$153.23
WHEATON® MicroLiter 11 mm Crimp-Top Seals, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$48.17
SureSTART™ Caps for 2 ml Glass Snap Top Vials, Level 2 High-Throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$461.12 to $461.12
Cap, 12mm solid top storage, 13-425 Closure screw Thread, Color: Black, Material: Polypropylene
Fournisseur: Thermo Scientific
$171.81
VWR® 11 mm Crimp Top Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$291.56
SureSTART™ Crimp Caps, 20 mm, Level 2 High-throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$180.01 to $182.41