Avantor® Hichrom, Inlet Solvent Filters
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
These solvent inlet filters are used at the low-pressure inlet side of the pump to help protect check valves, injector and column from particulate contamination from the eluent.
- Helps to remove dissolved gases
The large surface area filters out particulate matter from the solvent that may otherwise damage expensive hardware. No tools are required for replacement.
Customers Who Bought This Also Bought