Avantor® Hichrom, PEEK Universal Column Coupler
# de catalogue 76399-136
Fournisseur: HICHROM LIMITED
Avantor® Hichrom, PEEK Universal Column Coupler
# de catalogue 76399-136
Fournisseur: HICHROM LIMITED
Spécifications
- I.D.:0.13 mm
- Description:Universal column coupler red
- Material:PEEK
- Cat. no.:76399-136
Spécifications
A propos de cet article
Customers Who Bought This Also Bought
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
63,72 $ to 462,44 $
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Fournisseur: VWR International
57,66 $ to 576,72 $
VWR® 11 mm Crimp Top Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
279,61 $
WHEATON® MicroLiter 20 mm Crimp Seals with Septa, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
196,79 $
SureSTART™ Crimp Caps, 20 mm, Level 2 High-throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
11 110,75 $ to 11 110,75 $
WHEATON® MicroLiter Sample Vial Component Kits, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
467,43 $