Avantor® Hichrom, Chromatography Tubing, Precut SS
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
316 Stainless steel, precision bore ¹/₁₆" type tubing.
- Passivated for chemical resistance
- Square, burr-free ends
- Precut lengths
0.005 and 0.007" Int. Ø - suitable for minimal dead volume injector-column or column-detector connections of microbore or short columns containing 3 µm material. Sample and material must be filtered to avoid blockage.
0.010" Int. Ø - suitable for connection of analytical and semi-preparative columns containing 5 and 10 µm material. It should be used in all parts of the system where dead volume needs to be kept to minimum.
0.20" Int. Ø - suitable for use in preparative systems or in non-critical connections (pump to sample injector where low flow resistance is more important than low internal volume).
Ext. Ø: 1.5 mm.
Customers Who Bought This Also Bought