Avantor® Hichrom, Connectors
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
The 76399-038 female-female connector consists of a 60 mm length of ¹/₁₆" (0.007 mm) PEEK tubing with PEEK fingertight fittings and is suitable for connecting to all end fittings.
The 76399-034 male-male connector provides an ultra low dead volume and comes complete with two PEEK end fittings.
- Low dead volume
- Male-male and female-female options
- Inert and biocompatible
Customers Who Bought This Also Bought