VeriSpec® ICP Multi-Element Internal Calibration Standard 7, RICCA Chemical Company
Fournisseur: Ricca Chemical
Certificats
A propos de cet article
Seven component standard.
10 ppm: ⁶Li, Sc, Y, In, Tb, Bi, Ge.
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.