VeriSpec® ICP Multi-Element Interference Check Solution 12, RICCA Chemical Company
Fournisseur: Ricca Chemical
Certificats
A propos de cet article
12 component standard.
10000 ppm: Cl
2000 ppm: C
1000 ppm: Al, Ca, Fe, K, Mg, Na, P, S
20 ppm: Mo, Ti
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.