SIM Lipoma Surgical Models
Fournisseur: SIMULATED INANIMATE MODELS LLC SE
Certificats
A propos de cet article
A single use surgical simulator with a real tissue feel.
- Made of water based hydrogel
- Imaging compatible
- Excellent tissue characteristics
SIM prostate phantoms are composed of a water-based hydrogel material that enables superior ultrasound imaging properties versus the competition and are compatible with both standard side-fire and end-fire ultrasound probes. These trainers are ideal for training in Lipoma diagnosis and removal. Each trainer contains a large lipoma for removal, it can be palpated and moved, biopsied, and surgically removed. The advanced trainer also allows for imaging tests prior to removal.
Packaging: For greatest shelf life, refrigerate on arrival, and leave sealed until used.
Customers Who Bought This Also Bought