A propos de cet article
Designed with ergonomics and intuitive handling in mind, HF bottle top dispenser helps in dispensing Hydrofluoric Acid with safety and precision.
- Springless Valve™ for smooth and jam-free functioning
- PTFE piston with ETP O-Ring for effortless piston movement and high chemical compatibility
- Easy and effortless volume setting
- Flexible delivery nozzle to change angle of dispensing
- Dispensers is fully autoclavable at 121 °C and 15 psi for a duration of 10 to 15 minutes
- Reproducibility of 0.2%
HF bottle top dispenser is designed with selected materials for barrel and valve assembly that enable the dispenser to be used with Hydrofluoric Acid (HF) in addition to solvents and other reagents.
Designed with ergonomics and intuitive handling in mind, HF bottle top dispenser exhibits excellent chemical compatibility and helps in achieving precision with reliability in practical laboratory environments. This dispenser uses a re-circulation valve which prevents the loss of reagents during purging by re-directing them into the mounted bottle, thus facilitating bubble free dispensing.
A specially designed volume adjustment knob of this dispenser allows 180° rotation for easy and effortless volume setting.