Buffers, Precision Reference Standards, RICCA Chemical Company
Supplier: Ricca Chemical
Certificates
About this item
This precision line of pH Buffers offers tolerances of ±0.005 pH unit or better.
Certifications: Compared against and certified traceable to NIST Standard Reference Materials.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.