Aluminum Forms Holders, Labelmaster
# de catalogue 101378-786
Fournisseur: Label Master
Spécifications
- Description:Holder for 8×14" forms
- Material:Recycled aluminum
- Cat. no.:101378-786
- Supplier No.:FH11019
Spécifications
A propos de cet article
Protect forms and transport them more easily with Aluminum Forms Holders.
- Heavy-duty recycled aluminum construction
- Clipboard and large writing surface
- Latched lid protects shipping forms, inspections forms, and logbooks
The top-mounted clip holds up to 1" of paper and includes a slide-on credit card and license holder. The holder features a 1.5" deep storage compartment with latched lid. It has a weather-resistant lacquer finish and a handy, built-in pen tray.
Customers Who Bought This Also Bought