Non-Hazardous Waste Manifest Form, Labelmaster
# de catalogue 101377-172
Fournisseur: Label Master
Spécifications
- Description:6-part snap-out form rev 3/95
- Height:11"
- Material:PP/cellulose
- Cat. no.:101377-172
- Width:8.5"
- Supplier No.:F14
Spécifications
A propos de cet article
Clearly identify non-hazardous waste with this non-hazardous waste manifest form.
- Available in multiple formats
The form minimizes the likelihood that non-hazardous waste will be mistaken for hazardous waste and helps ensure that non-hazardous materials are not classified as hazardous materials when both are contained in the same shipment. Use Masterform software to fill out the laser form.
Packaging: Sold in packages of 100 sheets.
Customers Who Bought This Also Bought