DOT Labels 'Explosive Class 1.4 S', Labelmaster
# de catalogue 101376-466
Fournisseur: Label Master
Spécifications
- Description:Preprinted dot labels
- Material:Semi-gloss paper
- Thickness:5.85 mil
- Cat. no.:101376-466
- Packaging:On roll
- Core Size:3"
- Legend:Explosive 1.4 S
- Dimensions:100×100 mm (3.94×3.94")
- Supplier No.:LEXP14S
Spécifications
A propos de cet article
Help stay in compliance with standards set by the DOT with Explosive Class 1.4 S labels, revised to meet the design requirements of 49 CFR 172.407.
- Printed with light-fast inks for fade resistance
- Adhesives formulated for use in various climates and environments
- Strong holds on corrugated boxes
- Once affixed, labels won't fade or fall off
- Outstanding chemcial and abrasion resistance
These labels also meet the design and durability standards of 49 CFR, ICAO, and other international air regulations.
Customers Who Bought This Also Bought