Labels 'Empty', Labelmaster
# de catalogue 101378-490
Fournisseur: Label Master
Spécifications
- Description:Preprinted labels
- Material:PVC-free film
- Thickness:7.1 mil
- Cat. no.:101378-490
- Packaging:On roll
- Legend:Empty
- Dimensions:152×151.9 mm (5.98×5.98")
- Supplier No.:SL18
Spécifications
A propos de cet article
Help put safety first with Empty labels. These are used for marking a container of radioactive materials.
- Made of paper
- Outstanding chemical and abrasion resistance
- Strong adhesion to corrugated boxes
Customers Who Bought This Also Bought