International Labels 'Explosive Class 1', Labelmaster
# de catalogue 101377-452
Fournisseur: Label Master
Spécifications
- Description:International preprinted labels
- Material:Permanent vinyl
- Thickness:11.2 mil
- Cat. no.:101377-452
- Packaging:On roll
- Core Size:3"
- Legend:Explosive 1
- Dimensions:100×100 mm (3.94×3.94")
- Supplier No.:SLEX32
Spécifications
A propos de cet article
Help meet the design and durability standards of 49 CFR, ICAO, and other international air regulations with vinyl International Wordless Explosive Class 1 labels, revised to meet the design requirements of 49 CFR 172.407.
- Printed with light-fast inks for fade resistance
- Adhesives formulated for use in various climates and environments
- Will not fade or fall off
Featuring a tab that includes all the compatibility letters to meet diverse needs, these labels help users stay in compliance with standards set forth by the DOT. Constructed of rugged, flexible material, they feature strong adhesion to painted, steel, fiber, and plastic surfaces. They will pass the 90-day IMO seawater immersion test.
Customers Who Bought This Also Bought