Radioactive III Labels, Labelmaster
# de catalogue 101377-916
Fournisseur: Label Master
Spécifications
- Description:Warning labels
- Material:PVC-free film
- Thickness:7.1 mil
- Cat. no.:101377-916
- Core Size:3"
- Legend:Radioactive III
- Dimensions:100×100 mm (3.94×3.94")
- Supplier No.:HMSL16
Spécifications
A propos de cet article
Employ a fool-proof shipping label solution with Worded Radioactive III labels, revised to meet the design requirements of 49 CFR 172.407.
- Printed with light-fast inks for fade resistance
- Adhesives are formulated for use in various climates and environments
- Strong holds on corrugated boxes
- Once affixed, labels won't fade or fall off
- Outstanding chemical and abrasion resistance
In addition to complying with directives set by the DOT, these labels also meet the design and durability standards of 49 CFR, ICAO, and other international air regulations.
Customers Who Bought This Also Bought