Labels 'Flammable Liquid', Labelmaster
# de catalogue 101378-390
Fournisseur: Label Master
Spécifications
- Description:Preprinted labels
- Material:Semi-gloss paper
- Thickness:5.85 mil
- Cat. no.:101378-390
- Packaging:On roll
- Legend:Flammable Liquid
- Dimensions:100×101.6 mm (3.94×4")
- Supplier No.:HML6C
Spécifications
A propos de cet article
Employ a fool-proof shipping label solution with Worded Flammable Liquid labels, revised to meet the design requirements of 49 CFR 172.407.
- Printed with light-fast inks for fade resistance
- Adhesives formulated for use in various climates and environments
- Strong holds on corrugated boxes
- Once affixed, labels won't fade or fall off
- Outstanding chemical and abrasion resistance
In addition to complying with directives set by the DOT, these labels also meet the design and durability standards of 49 CFR, ICAO, and other international air regulations.
Customers Who Bought This Also Bought