Brown Kraft Paper Labels, Blank, Labelmaster
# de catalogue 101376-138
Fournisseur: Label Master
Spécifications
- Description:Brown kraft paper labels, blank
- Material:Brown kraft
- Thickness:6 mil
- Cat. no.:101376-138
- Dimensions:76.2×76.2 mm (3×3")
- Supplier No.:SHIPBR3
Spécifications
A propos de cet article
Explore labeling possibilities with Blank Brown Kraft Paper labels.
- Outstanding chemical and abrasion resistance
- Strong adhesion to corrugated boxes
These labels are perfect for covering up incorrect information, enabling users to reuse boxes.
Customers Who Bought This Also Bought