Labels 'Hazardous Waste', Labelmaster
Fournisseur: Label Master
Certificats
A propos de cet article
Avoid potential dangers and stay in regulatory compliance with Hazardous Waste labels.
- Constructed of paper
- Economical alternative to vinyl
These labels provide a convenient way for a generator to mark accumulation containers in accordance with EPA regulations in 40 CFR part 262.34.
Customers Who Bought This Also Bought