RQ Labels, Labelmaster
# de catalogue 101376-268
Fournisseur: Label Master
Spécifications
- Description:ORM RQ labels
- Material:Semi-gloss paper
- Thickness:5.85 mil
- Cat. no.:101376-268
- Legend:RQ
- Supplier No.:RQM
- Diameter:1.4375"
Spécifications
A propos de cet article
Ship hazardous substances with confidence when using RQ labels.
- Made of paper
- Outstanding chemical and abrasion resistance
- Strong adhesion to corrugated boxes
These labels are specifically used to satisfy the hazardous substance marking requirements of 172.324(b).
Customers Who Bought This Also Bought