Radioactive I Labels, Labelmaster
# de catalogue 101376-364
Fournisseur: Label Master
Spécifications
- Description:Warning labels
- Material:Semi-gloss paper
- Thickness:5.85 mil
- Cat. no.:101376-364
- Core Size:3"
- Legend:Radioactive I
- Dimensions:100×100 mm (3.94×3.94")
- Supplier No.:HML14
Spécifications
A propos de cet article
Ship with confidence when using Worded Radioactive I labels, revised to meet the design requirements of 49 CFR 172.407.
- Printed with light-fast inks for fade resistance
- Adhesives are formulated for use in various climates and environments
- Once affixed, labels won't fade or fall off
- Outstanding chemical and abrasion resistance
- White and black
In addition to complying with directives set by the DOT, these labels also meet the design and durability standards of 49 CFR, ICAO, and other international air regulations.
Customers Who Bought This Also Bought