Black Arrows Up Labels, Labelmaster
# de catalogue 101376-242
Fournisseur: Label Master
Spécifications
- Description:Arrows labels
- Material:Semi-gloss paper
- Thickness:5.85 mil
- Cat. no.:101376-242
- Legend:Black Arrows Up
- Dimensions:114.3×82.6 mm (4.5×3.25")
- Supplier No.:L22
Spécifications
A propos de cet article
Protect products from mishandling during shipment with Black Arrows Up labels.
- Printed on semi-gloss paper
- Outstanding chemical and abrasion resistance
- Strong adhesion to corrugated boxes
- Helps to maintain regulatory compliance
These labels comply with the requirements of 49 CFR 172.312 for marking the handling orientation on a hazardous materials package.
Customers Who Bought This Also Bought