Labels 'Non-RCRA Regulated Waste', Labelmaster
Fournisseur: Label Master
Certificats
A propos de cet article
Play it safe when shipping non-RCRA regulated waste with Non-RCRA Regulated Waste labels.
- Constructed of paper
- Economical alternative to vinyl
Using a PC and laser printer, the label's unruled lines allow users to fill in the information necessary for marking a container for transport in compliance with the DOT and EPA regulations.
Customers Who Bought This Also Bought