Labels 'Empty', Labelmaster
# de catalogue 101376-812
Fournisseur: Label Master
Spécifications
- Description:Preprinted labels
- Material:PVC-free film
- Thickness:7.1 mil
- Cat. no.:101376-812
- Core Size:3"
- Legend:Empty
- Dimensions:152.4×152.4 mm (6×6")
- Supplier No.:WMTV
Spécifications
A propos de cet article
Maintain the safety of your personnel when working with empty drums by using empty labels.
- Used to identify containers that previously contained hazardous materials
- Constructed of rugged and flexible PVC-free vinyl
- White and blue
These labels can be used to identify containers that previously contained hazardous materials. Using a PC and printer, the ruled lines allow users to fill in the information necessary for marking a container for transport in compliance with DOT and EPA regulations.
Customers Who Bought This Also Bought