Revised Organic Peroxide Labels, Labelmaster
# de catalogue 101376-392
Fournisseur: Label Master
Spécifications
- Description:Warning labels
- Material:Semi-gloss paper
- Thickness:5.85 mil
- Cat. no.:101376-392
- Core Size:3"
- Legend:Organic Peroxide
- Dimensions:100×100 mm (3.94×3.94")
- Supplier No.:HML52
Spécifications
A propos de cet article
Help take the guessing out of shipping label solutions with Revised Organic Peroxide labels.
- Once affixed, they won't fade or fall off
- Provide outstanding chemical and abrasion resistance
- Yellow and red with black type
These Hazard Class 5 labels are printed with light-fast inks for fade resistance, and adhesives are formulated for use in various climates and environments to ensure strong holds on corrugated boxes.
Certifications: Revised to meet the design requirements of 49 CFR 172.407. Complies with the directives of the Department of Transportation. Meets the design and durability standards of 49 CFR, ICAO, and other international air regulations.
Customers Who Bought This Also Bought