Wall Rack, 1×1 gallon, Marlin Steel Wire Products
# de catalogue 76338-196
Fournisseur: Marlin Steel Wire Products
Spécifications
- Description:Wall rack
- Material:Plain steel
- Cat. no.:76338-196
Spécifications
A propos de cet article
Powder coated steel 1x1 gallon wall rack.
- Frame diameter is 0.188"
- Crosswire diameter is 0.121"
Features red finish for high visibility and durable engineering, making it ideal for holding gallon-sized containers or bottles.
Customers Who Bought This Also Bought