Mesh Basket with Hinged Lid, Marlin Steel Wire Products
# de catalogue 76338-110
Fournisseur: Marlin Steel Wire Products
Spécifications
- Dimensions:11L×5.63W×2.77"H
- Description:Mesh basket with hinged lid
- Material:Stainless steel
- Cat. no.:76338-110
Spécifications
A propos de cet article
This stainless steel basket has a hinged lid to make sure your contents are secure.
- Electropolished finish
- With 12 mesh openings per linear inch
It is Ideal for storage and materials handling.
Customers Who Bought This Also Bought