Rectangular Basket, Marlin Steel Wire Products
# de catalogue 76338-088
Fournisseur: Marlin Steel Wire Products
Spécifications
- Dimensions:18L×18W×9"H
- Description:Rectangular basket
- Material:Stainless steel
- Cat. no.:76338-088
Spécifications
A propos de cet article
Rectangular material handling basket with handles and mesh
- Electropolished finish
- With four mesh openings per linear inch
- Handles for easy gripping
- Stacks so that parts inside the basket do not get crushed
Robust quality that is ideal for washing parts, parts degreasing, ultrasonic cleaning, curing, and materials handling in a variety of applications such as chemical bath, heat treating, and cryogenics.
Customers Who Bought This Also Bought