Crosswire Basket, Marlin Steel Wire Products
# de catalogue 76338-138
Fournisseur: Marlin Steel Wire Products
Spécifications
- Dimensions:20.25L×8W×6"H
- Description:Crosswire basket
- Material:Stainless steel 316
Spécifications
A propos de cet article
Stainless steel crosswire basket.
- Natural finish
- Deep dimensions allow for wide variety of uses
Ideal for many materials handling applications.
Customers Who Bought This Also Bought