Mesh Baskets, Rectangular, Marlin Steel Wire Products
Supplier: Marlin Steel Wire Products
Certificates
About this item
Rectangular material handling basket with handles and mesh.
- Electropolished finish
- With four mesh openings per linear inch
- Handles for easy handling between work stations
- Stacks so that parts inside the basket do not get crushed
This rectangular basket is compatible with a variety of equipment. Ideal for parts washing, degreasing, ultrasonic cleaning, curing, and material handling applications in extreme temperatures.
Customers Who Bought This Also Bought