Mesh Basket, Round, Marlin Steel Wire Products
# de catalogue 76338-066
Fournisseur: Marlin Steel Wire Products
Spécifications
- Dimensions:6.63D×6.63"H
- Description:Round mesh basket
- Material:Plain steel
- Cat. no.:76338-066
Spécifications
A propos de cet article
Round material handling basket with handle and mesh.
- Natural finish
- With four mesh openings per linear inch
- Handles for easy gripping
- Stacks so that parts inside the basket do not get crushed
Robust quality that is ideal for washing parts, cleaning, degreasing, ultrasonic cleaning, curing and materials handling in a variety of applications such as chemical baths, heat treating and cryogenics.
Customers Who Bought This Also Bought