Perforated Baskets, Round, Marlin Steel Wire Products
Fournisseur: Marlin Steel Wire Products
Certificats
A propos de cet article
Circular material handling basket with handles.
- Natural finish
- With four mesh openings per linear inch
- Handles for easy gripping
- Stacks so that parts inside the basket do not get crushed
Ideal for washing parts, parts degreasing, ultrasonic cleaning, curing, and materials handling in a variety of applications such as chemical bath, heat treating, and cryogenics.
Customers Who Bought This Also Bought