Mesh Baskets with Lid, Marlin Steel Wire Products
Fournisseur: Marlin Steel Wire Products
Certificats
A propos de cet article
Stainless steel material handling parts washing basket with a lid and hinge.
- Natural finish
- Multiple mesh sizes available
- Front latch on basket for secure closure
Used for small material handling or washing. Rugged construction ensures basket longevity in applications such as dipping, cleaning, or moving components in industrial settings.
Customers Who Bought This Also Bought