Material Handling Baskets, Round, Marlin Steel Wire Products
Fournisseur: Marlin Steel Wire Products
Certificats
A propos de cet article
Round material handling basket with handles.
- Natural finish
- With 24 mesh openings per linear inch
- Handles for easy gripping
- Stacks so that parts inside the basket do not get crushed
Robust quality that is ideal for washing parts, cleaning, degreasing, ultrasonic cleaning, curing, and materials handling in a variety of applications such as chemical baths, heat treating, and cryogenics.
Customers Who Bought This Also Bought