Expanded Metal Tote Baskets, Marlin Steel Wire Products
Fournisseur: Marlin Steel Wire Products
Certificats
A propos de cet article
Rectangular material handling basket with handles and mesh.
- Zinc plated or natural finish
- 0.25" or 0.50" mesh openings
- Handles for easy gripping
- Stacks so that parts inside the basket do not get crushed
Ideal for washing parts, parts degreasing, ultrasonic cleaning, curing, and materials handling in a variety of applications such as chemical bath, heat treating, and cryogenics.
Customers Who Bought This Also Bought